Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004364.2 | NP_001004364.2 | 7 | 950 | ||
NM_001098168.1 | NP_001091637.1 | 7 | 950 | |||
NM_001135192.1 | NP_001128664.1 | 7 | 950 | |||
GenBank mRNA | AK147622.1 | - | 7 | 958 | ||
AK147656.1 | - | 7 | 896 | |||
AK162242.1 | - | 249 | ||||
AK172944.1 | - | 7 | 762 | |||
BC080847.1 | - | 7 | 950 | 6852788 | ||
BC096022.1 | - | 7 | 718 | 30542525 | ||
BM944240.1 | - | 2 | 145 |