Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177600.4 | NP_808268.2 | 16 | 1905 | ||
GenBank mRNA | AK029862.1 | - | 8 | 1112 | ||
BC095976.1 | - | 15 | 1651 |