Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8030462N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178670.3 | NP_848785.2 | 2 | 794 | ||
GenBank mRNA | AK033209.1 | - | 2 | 781 | ||
AK046785.1 | - | 2 | 773 | |||
AK087799.1 | - | 2 | 718 | |||
AK139552.1 | - | 2 | 691 | |||
AK163840.1 | - | 795 | ||||
BC120889.2 | - | 1 | 504 | 40060928 | ||
DQ465969.1 | - | 1 | 197 | |||
DQ465977.1 | - | 197 | ||||
DQ465978.1 | - | 79 |