Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A730008H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172505.4 | NP_766093.1 | 5 | 1510 | ||
GenBank mRNA | AK042589.1 | - | 5 | 1510 | ||
AK138428.1 | - | 5 | 1473 | |||
AK190226.1 | - | 1 | 337 | |||
BC131919.1 | - | 5 | 1478 | 40130196 |
Gene Symbol: 6430706D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032612.1 | - | 2 | 812 | ||
AK135880.1 | - | 1 | 333 | |||
BC062125.1 | - | 5 | 1220 | 6826662 |
Gene Symbol: Hjurp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198652.2 | NP_941054.2 | 8 | 1759 | ||
GenBank mRNA | AB162219.1 | - | 9 | 1598 |