Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145381.2 | NP_663356.1 | 5 | 676 | ||
GenBank mRNA | AK088932.1 | - | 5 | 689 | ||
AK090091.1 | - | 5 | 706 | |||
AK143912.1 | - | 5 | 655 | |||
BC004045.1 | - | 5 | 676 | 3496398 |