Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164559.1 | NP_001158031.1 | 16 | 1934 | ||
NM_001164560.1 | NP_001158032.1 | 16 | 1858 | |||
NM_198020.2 | NP_932137.2 | 16 | 1961 | |||
GenBank mRNA | AK032945.1 | - | 14 | 3394 | ||
AK084775.1 | - | 16 | 1900 | |||
AK155251.1 | - | 16 | 1927 | |||
AK159467.1 | - | 16 | 1899 | |||
AK170412.1 | - | 16 | 1823 | |||
AK171002.1 | - | 16 | 1926 | |||
BB055133.2 | - | 4 | 489 | |||
BC027404.1 | - | 16 | 1930 | 4950980 |