Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172507.5 | NP_766095.1 | 2 | 270 | ||
GenBank mRNA | AK029392.1 | - | 259 | |||
AK032092.1 | - | 283 | ||||
AK076202.1 | - | 2 | 259 | |||
AK160357.1 | - | 2 | 259 | |||
BC038473.1 | - | 2 | 309 | 5008777 |