Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139146.2 | NP_631885.1 | 10 | 1965 | ||
GenBank mRNA | AF319623.1 | - | 10 | 1965 | ||
AJ606045.1 | - | 9 | 1683 | |||
AK129270.1 | - | 9 | 2956 | |||
BC138625.1 | - | 9 | 1716 | 8861645 | ||
BC138626.1 | - | 9 | 1716 | 8861646 |