Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl14ep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025102.1 | NP_001020273.1 | 3 | 645 | ||
NM_173750.2 | NP_776111.1 | 3 | 599 | |||
GenBank mRNA | AK076781.1 | - | 3 | 600 | ||
AK134965.1 | - | 3 | 636 | |||
BC054106.1 | - | 3 | 631 | 30021622 |