Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146690.1 | NP_001140162.1 | 7 | 1072 | ||
NM_144807.3 | NP_659056.3 | 7 | 1072 | |||
GenBank mRNA | AK007964.1 | - | 7 | 992 | ||
AK032497.1 | - | 7 | 967 | |||
AY445814.1 | - | 7 | 967 | |||
BE650891.1 | - | 3 | 307 |