Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172509.3 | NP_766097.1 | 4 | 1019 | ||
GenBank mRNA | AK029942.1 | - | 4 | 995 | ||
AK034583.1 | - | 4 | 995 | |||
AK083583.1 | - | 4 | 1020 | |||
AK140606.1 | - | 4 | 994 | |||
AK162568.1 | - | 4 | 1048 | |||
AK178565.1 | - | 1 | 138 | |||
AK195855.1 | - | 1 | 138 | |||
BC051544.1 | - | 3 | 562 | |||
BC139070.1 | - | 4 | 921 | 8862092 | ||
BC141201.1 | - | 4 | 921 | 9055813 |