Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172509.3 | NP_766097.1 | 2 | 484 | ||
GenBank mRNA | AK029942.1 | - | 2 | 459 | ||
AK034583.1 | - | 2 | 459 | |||
AK083583.1 | - | 2 | 485 | |||
AK140606.1 | - | 2 | 459 | |||
AK162568.1 | - | 2 | 512 | |||
BC051544.1 | - | 1 | 27 | |||
BC139070.1 | - | 2 | 386 | 8862092 | ||
BC141201.1 | - | 2 | 386 | 9055813 |