Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178079.3 | NP_835180.2 | 3 | 309 | ||
GenBank mRNA | AK028883.1 | - | 3 | 306 | ||
AK128969.1 | - | 3 | 315 | |||
AK143953.1 | - | 3 | 304 | |||
BC120725.1 | - | 3 | 309 | 40129648 | ||
BC125395.1 | - | 3 | 309 | 40129910 |