Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tifab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168615.1 | NP_001162086.1 | 3 | 882 | ||
NM_145976.4 | NP_666088.1 | 3 | 818 | |||
GenBank mRNA | AK036643.1 | - | 3 | 735 | ||
AK153654.1 | - | 3 | 614 | |||
AK170349.1 | - | 3 | 663 | |||
AK220206.1 | - | 1 | 369 | |||
BC005528.1 | - | 3 | 516 | 3495346 | ||
BC027057.1 | - | 2 | 401 | 5029696 | ||
BC031130.1 | - | 3 | 650 | 4954852 |