Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam46a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160378.1 | NP_001153850.1 | 3 | 1251 | ||
NM_001160379.1 | NP_001153851.1 | 3 | 1222 | |||
GenBank mRNA | AK135132.1 | - | 3 | 1225 | ||
BC023892.1 | - | 1 | 491 |