Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145387.4 | NP_663362.2 | 9 | 1470 | ||
GenBank mRNA | AK077662.1 | - | 9 | 1471 | ||
AK085600.1 | - | 8 | 1641 | |||
AK171694.1 | - | 8 | 1380 | |||
AK220240.1 | - | 5 | 2702 | |||
BC056953.1 | - | 9 | 1432 | 5717810 |