Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145387.4 | NP_663362.2 | 2 | 329 | ||
GenBank mRNA | AK077662.1 | - | 2 | 330 | ||
AK085600.1 | - | 2 | 345 | |||
AK220240.1 | - | 2 | 289 | |||
BC023151.1 | - | 1 | 15 | |||
BC056953.1 | - | 2 | 291 | 5717810 |