Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253736.1 | NP_001240665.1 | 8 | 794 | ||
NM_145978.2 | NP_666090.1 | 8 | 721 | |||
GenBank mRNA | BC024556.1 | - | 8 | 721 | 4990983 | |
BE634101.1 | - | 4 | 216 |