Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175440.4 | NP_780649.1 | 4, 5 | 583 | ||
GenBank mRNA | AF542056.1 | - | 4, 5 | 519 | ||
AK080281.1 | - | 4, 5 | 584 | |||
BC117009.1 | - | 4, 5 | 566 | 40126328 | ||
BC117011.1 | - | 4, 5 | 566 | 40126330 |