Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153762.3 | NP_717095.2 | 1 | 917 | ||
GenBank mRNA | AK033164.1 | - | 1 | 1371 | ||
AK085078.1 | - | 1367 | ||||
AK157821.1 | - | 1 | 1366 | |||
BC004739.1 | - | 1 | 968 | 3501511 | ||
BC063251.1 | - | 1 | 917 | 6413980 | ||
BC111036.1 | - | 2 | 906 |