Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178045.4 | NP_835146.3 | 7 | 767 | ||
GenBank mRNA | AK036528.1 | - | 7 | 731 | ||
AK089460.1 | - | 7 | 725 | |||
AK139284.1 | - | 7 | 714 | |||
AK151561.1 | - | 7 | 732 | |||
AK152639.1 | - | 7 | 732 | |||
AK160584.1 | - | 7 | 741 | |||
AK164357.1 | - | 7 | 730 | |||
AK170955.1 | - | 7 | 716 | |||
AK171198.1 | - | 7 | 745 | |||
BC060709.1 | - | 7 | 722 | 6409692 |