Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A630033H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122595.1 | NP_001116067.1 | 2 | 1008 | ||
NM_001122596.1 | NP_001116068.1 | 2 | 1055 | |||
NM_175442.4 | NP_780651.1 | 2 | 1019 | |||
GenBank mRNA | AK041740.1 | - | 2 | 1020 |