Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172516.4 | NP_766104.2 | 7 | 2020 | ||
GenBank mRNA | AK043199.1 | - | 2 | 889 | ||
AK050542.1 | - | 3 | 266 | |||
AK083696.1 | - | 7 | 1918 | |||
AK132119.1 | - | 7 | 1895 | |||
AK145449.1 | - | 7 | 1865 | |||
AK172954.1 | - | 7 | 1866 | |||
AK183448.1 | - | 3 | 305 | |||
AK184349.1 | - | 3 | 305 | |||
AY208852.1 | - | 7 | 1867 | |||
AY429676.1 | - | 7 | 1906 | |||
BC019133.1 | - | 5 | 655 | |||
BC089380.1 | - | 4 | 437 |