Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153136.4 | NP_694776.2 | 2 | 277 | ||
GenBank mRNA | AK137014.1 | - | 2 | 488 | ||
AK145997.1 | - | 2 | 266 | |||
AK155086.1 | - | 2 | 278 | |||
AK157937.1 | - | 2 | 278 | |||
BC036718.1 | - | 2 | 433 | 5322150 |