Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033253.3 | NP_001028425.1 | 16 | 3455 | ||
NM_001159942.1 | NP_001153414.1 | 17 | 3621 | |||
GenBank mRNA | AK122468.1 | - | 13 | 2779 | ||
AK135923.1 | - | 16 | 3457 | |||
BC076628.1 | - | 15 | 2817 | |||
BC117965.1 | - | 15 | 3057 | 40098326 | ||
BC117966.1 | - | 15 | 3050 | 40098330 |