Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144812.2 | NP_659061.2 | 13 | 3743 | ||
NM_177124.4 | NP_796098.3 | 10 | 3534 | |||
GenBank mRNA | AK052247.1 | - | 5 | 497 | ||
AK162702.1 | - | 4 | 234 | |||
AK162788.1 | - | 5 | 790 | |||
BC068158.1 | - | 10 | 3429 | |||
BC115729.1 | - | 6 | 827 |