Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134399.1 | NP_001127871.1 | 9 | 1177 | ||
NM_172522.4 | NP_766110.3 | 8 | 1084 | |||
GenBank mRNA | AK032661.1 | - | 8 | 896 | ||
AK051642.1 | - | 8 | 1021 | |||
AK122555.2 | - | 9 | 1117 | |||
BC151157.1 | - | 9 | 1078 | 9088045 | ||
BC172024.1 | - | 9 | 1078 | 9054310 |