Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172338.2 | NP_758841.1 | 13, 14 | 1851 | ||
GenBank mRNA | AK028692.1 | - | 13, 14 | 1856 | ||
AK051048.1 | - | 13, 14 | 1852 | |||
AK122407.1 | - | 2098 | ||||
AK154946.1 | - | 1872 | ||||
AK164023.1 | - | 13, 14 | 1867 | |||
BC046424.1 | - | 7, 8 | 846 | 3989412 |