Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172338.2 | NP_758841.1 | 15 | 2178 | ||
GenBank mRNA | AK028692.1 | - | 15 | 2183 | ||
AK051048.1 | - | 15 | 2179 | |||
AK122407.1 | - | 2425 | ||||
AK154946.1 | - | 2199 | ||||
AK164023.1 | - | 15 | 2194 | |||
BC046424.1 | - | 9 | 1173 | 3989412 |