Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040400.2 | NP_001035490.2 | 5658 | |||
GenBank mRNA | AK035157.1 | - | 1 | 37 | ||
AK129389.1 | - | 4 | 2845 | |||
AK152297.1 | - | 9 | 2648 | |||
AK153251.1 | - | 9 | 2648 | |||
AK153311.1 | - | 9 | 2648 | |||
AK153460.1 | - | 9 | 2648 | |||
BC031159.1 | - | 3 | 1833 | |||
BC040785.1 | - | 3 | 1833 | |||
DQ079067.1 | - | 12 | 5657 | |||
GU079949.1 | - | 10 | 5303 |