Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004564.2 | NP_004555.1 | 2, 3 | 366 | ||
XM_005263063.3 | XP_005263120.1 | 2, 3 | 328 | |||
GenBank mRNA | AB019410.2 | - | 2, 3 | 310 | ||
AF026851.1 | - | 2, 3 | 337 | |||
AF151033.1 | - | 2, 3 | 334 | |||
AK222887.1 | - | 2, 3 | 328 | |||
AK225241.1 | - | 2, 3 | 328 | |||
AK303461.1 | - | 2, 3 | 313 | |||
AK312957.1 | - | 2, 3 | 328 | |||
BC130348.1 | - | 2, 3 | 376 | 40146379 | ||
BC136547.1 | - | 2, 3 | 376 | 9020537 | ||
BP348031.1 | - | 2, 3 | 366 |