Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172526.3 | NP_766114.1 | 9 | 1524 | ||
GenBank mRNA | AK039884.1 | - | 9 | 1525 | ||
AK076743.1 | - | 4 | 340 | |||
BC055114.1 | - | 4 | 339 | |||
BC147554.1 | - | 8 | 1228 | 9056545 | ||
BC147555.1 | - | 8 | 1228 | 9056546 |