Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175443.5 | NP_780652.2 | 4 | 942 | ||
GenBank mRNA | AK031665.1 | - | 4 | 728 | ||
AK077179.1 | - | 4 | 681 | |||
AK087818.1 | - | 3 | 740 | |||
AY212244.1 | - | 4 | 533 | |||
BC145351.1 | - | 4 | 721 | 9053907 | ||
BC152310.1 | - | 4 | 880 | 40089976 |