Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153538.3 | NP_705766.3 | 20 | 3594 | ||
GenBank mRNA | AK031043.1 | - | 19 | 3171 | ||
AK162319.1 | - | 5 | 374 | |||
BC023438.1 | - | 8 | 947 | |||
BC023880.1 | - | 12 | 2305 | 5354757 | ||
BC025485.1 | - | 8 | 1729 | |||
BC043111.1 | - | 20 | 3548 | 6407010 | ||
CB526652.1 | - | 2 | 125 |