Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044380.1 | NP_001037845.1 | 5 | 1580 | ||
GenBank mRNA | AK129450.1 | - | 975 | |||
AK138646.1 | - | 5 | 1304 | |||
AK147996.1 | - | 1247 | ||||
AK166269.1 | - | 1476 | ||||
BC057108.1 | - | 573 | ||||
BC113163.1 | - | 5 | 1580 | 40060054 | ||
BC113773.1 | - | 5 | 1576 | 40060051 |