Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146048.1 | NP_001139520.1 | 12 | 1351 | ||
NM_172528.3 | NP_766116.3 | 12 | 999 | |||
GenBank mRNA | AK082721.1 | - | 12 | 1000 | ||
AK147075.1 | - | 12 | 1351 | |||
AK220166.1 | - | 12 | 1369 | |||
BC022613.1 | - | 4 | 293 | |||
BC046591.1 | - | 12 | 1338 | |||
BC087542.1 | - | 12 | 1198 | 4934980 |