Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145991.1 | NP_666103.1 | 7 | 655 | ||
GenBank mRNA | AK080861.1 | - | 7 | 787 | ||
BC027756.1 | - | 7 | 756 | 5067496 | ||
BC031127.1 | - | 7 | 655 | 4972826 |