Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134741.1 | NP_001128213.1 | 15 | 2497 | ||
NM_001277730.1 | NP_001264659.1 | 15 | 2203 | |||
GenBank mRNA | AK132674.1 | - | 15 | 2482 | ||
AK133026.1 | - | 13 | 1870 | |||
AY522557.1 | - | 9 | 1357 | |||
BC099972.1 | - | 15 | 2203 | 30080156 |