Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172532.3 | NP_766120.1 | 10 | 1624 | ||
GenBank mRNA | AK052703.1 | - | 10 | 1625 | ||
AK144030.1 | - | 10 | 1608 | |||
AK159485.1 | - | 10 | 1604 | |||
AK166313.1 | - | 10 | 1604 | |||
BC138703.1 | - | 10 | 1587 | 8861725 |