Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039534.1 | NP_001034623.1 | 6 | 1003 | ||
GenBank mRNA | AK077585.1 | - | 6 | 993 | ||
AU255311.1 | - | 1 | 21 | |||
AV333054.1 | - | 1 | 75 | |||
BC030313.1 | - | 4 | 1495 | |||
BC110381.1 | - | 6 | 979 | 5149569 |