Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000288.3 | NP_000279.1 | 8, 9 | 909 | ||
XM_005267019.4 | XP_005267076.1 | 8, 9 | 1189 | |||
XM_006715502.1 | XP_006715565.1 | 5, 6 | 959 | |||
GenBank mRNA | BC006268.1 | - | 8, 9 | 851 | 3948721 | |
BC031606.1 | - | 9, 10 | 1001 | 5176358 | ||
CR542054.1 | - | 8, 9 | 807 | |||
CR542076.1 | - | 8, 9 | 807 | |||
U76560.1 | - | 8, 9 | 855 | |||
U88871.1 | - | 8, 9 | 888 |