Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 2, 3 | 649 | ||
NM_172257.4 | NP_758461.1 | 2, 3 | 649 | |||
GenBank mRNA | AK081177.1 | - | 2, 3 | 635 | ||
AK152371.1 | - | 2, 3 | 582 | |||
AK155875.1 | - | 2, 3 | 553 | |||
AK196459.1 | - | 2, 3 | 181 | |||
BC023957.1 | - | 2, 3 | 653 | 5321190 | ||
BC051101.1 | - | 2, 3 | 377 | 5052849 | ||
CB247121.1 | - | 2, 3 | 658 |