Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081351.1 | NP_001074820.1 | 3 | 1429 | ||
GenBank mRNA | AK032959.1 | - | 2 | 116 | ||
BC151018.1 | - | 3 | 944 | 9087931 |