Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002617.3 | NP_002608.1 | 3 | 609 | ||
NM_153818.1 | NP_722540.1 | 3 | 609 | |||
XM_011541573.1 | XP_011539875.1 | 3 | 660 | |||
XM_011541576.1 | XP_011539878.1 | 3 | 660 | |||
GenBank mRNA | AB013818.1 | - | 3 | 609 | ||
AF060502.1 | - | 3 | 572 | |||
AK124816.1 | - | 3 | 587 | |||
BC000543.2 | - | 3 | 568 | 3163782 | ||
BC018198.1 | - | 3 | 591 | 3882695 | ||
CR542077.1 | - | 3 | 540 |