Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039676.2 | NP_001034765.2 | 3 | 454 | ||
GenBank mRNA | AK089322.1 | - | 3 | 406 | ||
AK135117.1 | - | 3 | 1171 | |||
AK170178.1 | - | 3 | 409 | |||
BC113769.1 | - | 3 | 424 | 40057862 |