Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039676.2 | NP_001034765.2 | 1 | 275 | ||
GenBank mRNA | AK089322.1 | - | 1 | 227 | ||
AK135117.1 | - | 1 | 991 | |||
AK170178.1 | - | 1 | 230 | |||
BC113769.1 | - | 1 | 245 | 40057862 | ||
BY200549.1 | - | 1 | 277 |