Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145999.2 | NP_666111.1 | 3 | 582 | ||
GenBank mRNA | AK142183.1 | - | 2 | 278 | ||
AY311390.1 | - | 3 | 582 | |||
BC029777.1 | - | 3 | 215 | 4950061 | ||
BC034062.1 | - | 3 | 237 | 4207654 |