Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290997.1 | NP_001277926.1 | 21 | 3111 | ||
NM_001291000.1 | NP_001277929.1 | 19 | 3442 | |||
NM_172537.4 | NP_766125.2 | 17 | 3268 | |||
NM_199238.3 | NP_954708.1 | 17 | 3229 | |||
NM_199239.3 | NP_954709.1 | 17 | 2560 | |||
NM_199240.3 | NP_954710.1 | 17 | 2632 | |||
NM_199241.3 | NP_954711.1 | 18 | 2689 | |||
GenBank mRNA | AB091532.1 | - | 17 | 3268 | ||
AB091533.1 | - | 16 | 2410 | |||
AB091534.1 | - | 17 | 2506 | |||
AB091535.1 | - | 17 | 2578 | |||
AB091536.1 | - | 18 | 2635 | |||
AK031307.1 | - | 19 | 3138 | |||
AK084029.1 | - | 1 | 59 | |||
AK122515.1 | - | 21 | 3114 | |||
BC060680.1 | - | 17 | 2523 | 6406252 |