Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172156.2 | NP_742168.1 | 247 | |||
GenBank mRNA | AF525161.1 | - | 247 | |||
BC068253.1 | - | 244 |
Gene Symbol: Prl3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205322.1 | NP_001192251.1 | 3 | 276 | ||
NM_008864.3 | NP_032890.2 | 3 | 273 | |||
GenBank mRNA | AF525162.1 | - | 247 | |||
AK082929.1 | - | 3 | 272 | |||
BC064736.1 | - | 3 | 256 | 30437709 | ||
M35662.1 | - | 3 | 253 |