Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300957.1 | NP_001287886.1 | 3 | 1310 | ||
NM_153786.2 | NP_722481.1 | 3 | 1313 | |||
GenBank mRNA | AF542181.1 | - | 3 | 1129 | ||
AJ578054.1 | - | 3 | 1131 | |||
AK048390.1 | - | 3 | 1285 | |||
AK052987.1 | - | 3 | 1285 | |||
BC133696.1 | - | 4 | 649 | 40081288 |